1,5-Dimethyl-1H-pyrazole-4-carbonitrile

Supplier: Matrix Scientific

077344-500MG 077344-1G 077344-5G
102665-706EA 646.15 USD
102665-706 102665-708 102665-710
1,5-Dimethyl-1H-pyrazole-4-carbonitrile
1,5-Dimethyl-1H-pyrazole-4-carbonitrile

Hazard: Irritant

Formula: C₆H₇N₃
MDL Number: MFCD04969708
CAS Number: 856860-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR