1-Methyl-1H-pyrazole-5-carbonitrile

Supplier: Matrix Scientific

077343-500MG 077343-1G 077343-5G
102665-700EA 516.92 USD
102665-700 102665-702 102665-704
1-Methyl-1H-pyrazole-5-carbonitrile
1-Methyl-1H-pyrazole-5-carbonitrile

Hazard: Irritant

Formula: C₅H₅N₃
MDL Number: MFCD09701708
CAS Number: 66121-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR