2-(3,4,5-Trimethyl-1H-pyrazol-1-yl)ethanol
Supplier: Matrix Scientific
077342-500MG
077342-1G
077342-5G
102665-694EA
387.69
USD
102665-694
102665-696
102665-698
2-(3,4,5-Trimethyl-1H-pyrazol-1-yl)ethanol
2-(3,4,5-Trimethyl-1H-pyrazol-1-yl)ethanol
Hazard: Irritant
Formula:
C₈H₁₄N₂O |
MDL Number:
MFCD14586864 CAS Number: 127223-53-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...