1,3-Dimethyl-1H-pyrazole-4-carbonitrile

Supplier: Matrix Scientific

077341-500MG 077341-1G
102665-690EA 516.92 USD
102665-690 102665-692
1,3-Dimethyl-1H-pyrazole-4-carbonitrile
1,3-Dimethyl-1H-pyrazole-4-carbonitrile

Hazard: Irritant

Formula: C₆H₇N₃
MDL Number: MFCD04969706
CAS Number: 87412-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR