3-Methyl-4,5-dihydro-1H-pyrazole

Supplier: Matrix Scientific

077339-500MG 077339-1G 077339-5G
102665-678EA 258.46 USD
102665-678 102665-680 102665-682
3-Methyl-4,5-dihydro-1H-pyrazole
3-Methyl-4,5-dihydro-1H-pyrazole

Hazard: Irritant

Formula: C₄H₈N₂
MDL Number: MFCD11518898
CAS Number: 1911-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR