4-Anilino-4-methylpentan-2-one hydrochloride
Supplier: Matrix Scientific
077330-500MG
077330-1G
077330-5G
102665-640EA
161.54
USD
102665-640
102665-642
102665-644
4-Anilino-4-methylpentan-2-one hydrochloride
4-Anilino-4-methylpentan-2-one hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₈ClNO |
MDL Number:
MFCD00569224 CAS Number: 88187-84-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...