1-Phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine

Supplier: Matrix Scientific

077320-500MG 077320-1G 077320-5G
102665-586EA 516.92 USD
102665-586 102665-588 102665-590
1-Phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine
1-Phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine

Hazard: Irritant

Formula: C₁₆H₁₇N
MDL Number: MFCD01817308
CAS Number: 20390-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR