1,2,3,4-Tetrahydroacridin-9-amine

Supplier: Matrix Scientific

077318-500MG 077318-1G 077318-5G
102665-574EA 387.69 USD
102665-574 102665-576 102665-578
1,2,3,4-Tetrahydroacridin-9-amine
1,2,3,4-Tetrahydroacridin-9-amine

Hazard: Irritant

Formula: C₁₃H₁₄N₂
MW: 198.26 g/mol
MDL Number: MFCD00046923
CAS Number: 321-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR