5-Pyridin-2-yl-1,3,4-oxadiazol-2-amine

Supplier: Matrix Scientific

Synonyms: 5-(Pyridin-2-yl)-1,3,4-oxadiazol-2-amine

077315-500MG 077315-1G 077315-5G
102665-560EA 193.85 USD
102665-560 102665-562 102665-564
5-Pyridin-2-yl-1,3,4-oxadiazol-2-amine
5-Pyridin-2-yl-1,3,4-oxadiazol-2-amine

Hazard: Irritant

Formula: C₇H₆N₄O
MW: 162.15 g/mol
MDL Number: MFCD01710636
CAS Number: 5711-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR