3-(2-Oxoazepan-1-yl)propanoic acid

Supplier: Matrix Scientific

077314-500MG 077314-1G 077314-5G
102665-554EA 258.46 USD
102665-554 102665-556 102665-558
3-(2-Oxoazepan-1-yl)propanoic acid
3-(2-Oxoazepan-1-yl)propanoic acid

Hazard: Irritant

Formula: C₉H₁₅NO₃
MDL Number: MFCD08059810
CAS Number: 505026-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR