N-Benzyl-N-methyl-β-alanine

Supplier: Matrix Scientific

Synonyms: 3-[Benzyl(methyl)amino]propanoic acid

077303-500MG 077303-1G 077303-5G
102665-504EA 516.92 USD
102665-504 102665-506 102665-508
N-Benzyl-N-methyl-β-alanine
N-Benzyl-N-methyl-beta-alanine

Hazard: Irritant

Formula: C₁₁H₁₅NO₂
MW: 193.25 g/mol
MDL Number: MFCD09717730
CAS Number: 149692-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR