N-Benzyl-N-methylglycine

Supplier: Matrix Scientific

Synonyms: 2-(Benzyl(methyl)amino)acetic acid

077301-500MG 077301-1G 077301-5G
102665-494EA 387.69 USD
102665-494 102665-496 102665-498
N-Benzyl-N-methylglycine
N-Benzyl-N-methylglycine

Hazard: Irritant

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
MDL Number: MFCD09711201
CAS Number: 37429-48-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR