2-Piperidin-1-ylacetohydrazide

Supplier: Matrix Scientific

077299-500MG 077299-1G 077299-5G
102665-482EA 258.46 USD
102665-482 102665-484 102665-486
2-Piperidin-1-ylacetohydrazide
2-Piperidin-1-ylacetohydrazide

Hazard: Irritant

Formula: C₇H₁₅N₃O
Boiling Pt: 130 °C (2 Torr)
Melting Pt: 154…156 °C
MDL Number: MFCD02269933
CAS Number: 7408-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR