Methyl-3-pyrrolidin-1-ylpropanoate

Supplier: Matrix Scientific

077298-500MG 077298-1G 077298-5G
102665-476EA 387.69 USD
102665-476 102665-478 102665-480
Methyl-3-pyrrolidin-1-ylpropanoate
Methyl-3-pyrrolidin-1-ylpropanoate

Hazard: Irritant

Formula: C₈H₁₅NO₂
MDL Number: MFCD01695467
CAS Number: 22041-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR