1-Butyl-1H-benzimidazole-2-sulfonic acid

Supplier: Matrix Scientific

077289-500MG 077289-1G 077289-5G
102665-430EA 387.69 USD
102665-430 102665-432 102665-434
1-Butyl-1H-benzimidazole-2-sulfonic acid
1-Butyl-1H-benzimidazole-2-sulfonic acid

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₃S
MDL Number: MFCD00964522
CAS Number: 300707-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR