2-Cyclopropyl-1H-benzimidazole

Supplier: Matrix Scientific

077286-500MG 077286-1G 077286-5G
102665-412EA 387.69 USD
102665-412 102665-414 102665-416
2-Cyclopropyl-1H-benzimidazole
2-Cyclopropyl-1H-benzimidazole

Hazard: Irritant

Formula: C₁₀H₁₀N₂
MDL Number: MFCD01859172
CAS Number: 16405-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR