2-(2-Furyl)-1H-benzimidazole
Supplier: Matrix Scientific
077285-500MG
077285-1G
077285-5G
102665-406EA
193.85
USD
102665-406
102665-408
102665-410
2-(2-Furyl)-1H-benzimidazole
2-(2-Furyl)-1H-benzimidazole
Hazard: Irritant
Formula:
C₁₁H₈N₂O |
MDL Number:
MFCD00055496 CAS Number: 3878-19-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...