2-(Phenoxymethyl)-1H-benzimidazole

Supplier: Matrix Scientific

077284-500MG 077284-1G 077284-5G
102665-400EA 258.46 USD
102665-400 102665-402 102665-404
2-(Phenoxymethyl)-1H-benzimidazole
2-(Phenoxymethyl)-1H-benzimidazole

Hazard: Irritant

Formula: C₁₄H₁₂N₂O
MDL Number: MFCD00159968
CAS Number: 6637-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR