2-Methyl-2-morpholin-4-ylpropan-1-ol

Supplier: Matrix Scientific

Synonyms: 2-Methyl-2-morpholinopropan-1-ol

077281-500MG 077281-1G 077281-5G
102665-382EA 516.92 USD
102665-382 102665-384 102665-386
2-Methyl-2-morpholin-4-ylpropan-1-ol
2-Methyl-2-morpholin-4-ylpropan-1-ol

Hazard: Irritant

Formula: C₈H₁₇NO₂
MW: 159.23 g/mol
MDL Number: MFCD11053919
CAS Number: 858197-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR