N~1~,N~1~-Dimethyl-1-(2-methylphenyl)ethane-1,2-diamine

Supplier: Matrix Scientific

077277-500MG 077277-1G 077277-5G
102665-360EA 193.85 USD
102665-360 102665-362 102665-364
N~1~,N~1~-Dimethyl-1-(2-methylphenyl)ethane-1,2-diamine
N~1~,N~1~-Dimethyl-1-(2-methylphenyl)ethane-1,2-diamine

Hazard: Irritant

Formula: C₁₁H₁₈N₂
MDL Number: MFCD09261967
CAS Number: 928000-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR