3-Ethoxymethacrolein

Supplier: Matrix Scientific

077276-500MG 077276-1G
102665-356EA 161.54 USD
102665-356 102665-358
3-Ethoxymethacrolein
3-Ethoxymethacrolein

Hazard: Irritant

Formula: C₆H₁₀O₂
CAS Number: 42588-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR