1,1-Diethoxy-3-methylbutane

Supplier: Matrix Scientific

077274-500MG 077274-1G
102665-348EA 153.85 USD
102665-348 102665-350
1,1-Diethoxy-3-methylbutane
1,1-Diethoxy-3-methylbutane

TCA listed: T
Hazard: Irritant

Formula: C₉H₂₀O₂
Boiling Pt: 167…168 °C (750 Torr)
Density: 0.8356/20°C
MDL Number: MFCD00051787
CAS Number: 3842-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR