1,1-Diethoxypentane

Supplier: Matrix Scientific

077273-500MG 077273-1G
102665-344EA 161.54 USD
102665-344 102665-346
1,1-Diethoxypentane
1,1-Diethoxypentane

Hazard: Irritant

Formula: C₉H₂₀O₂
MDL Number: MFCD00129693
CAS Number: 3658-79-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR