2-Amino-5-methyl-1,3-thiazole-4-carboxylic acid

Supplier: Matrix Scientific

077271-500MG 077271-1G 077271-5G
102665-334EA 516.92 USD
102665-334 102665-336 102665-338
2-Amino-5-methyl-1,3-thiazole-4-carboxylic acid
2-Amino-5-methyl-1,3-thiazole-4-carboxylic acid

Hazard: Irritant

Formula: C₅H₆N₂O₂S
MDL Number: MFCD09991769
CAS Number: 688064-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR