4-Ethyl-1H-pyrazole

Supplier: Matrix Scientific

077266-500MG 077266-1G
102665-310EA 769.23 USD
102665-310 102665-312
4-Ethyl-1H-pyrazole
4-Ethyl-1H-pyrazole

Hazard: Irritant

Formula: C₅H₈N₂
MDL Number: MFCD18449289
CAS Number: 17072-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR