3-Phenyl-1,2,4-oxadiazol-5-ol

Supplier: Matrix Scientific

077263-500MG 077263-1G 077263-5G
102665-292EA 387.69 USD
102665-292 102665-294 102665-296
3-Phenyl-1,2,4-oxadiazol-5-ol
3-Phenyl-1,2,4-oxadiazol-5-ol

Hazard: Irritant

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
MDL Number: MFCD17676121
CAS Number: 1456-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR