5-Chloro-3-phenyl-1,2,4-oxadiazole

Supplier: Matrix Scientific

077262-500MG 077262-1G 077262-5G
102665-286EA 320 USD
102665-286 102665-288 102665-290
5-Chloro-3-phenyl-1,2,4-oxadiazole
5-Chloro-3-phenyl-1,2,4-oxadiazole

Hazard: Irritant

Formula: C₈H₅ClN₂O
MW: 180.59 g/mol
MDL Number: MFCD09971531
CAS Number: 827-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR