{[3-(4-Methylphenyl)-1,2,4-oxadiazol-5-yl]methyl}amine hydrochloride

Supplier: Matrix Scientific

Synonyms: [3-(4-Methylphenyl)-1,2,4-oxadiazol-5-yl]methanamine

077261-500MG 077261-1G
102665-282EA 387.69 USD
102665-282 102665-284
{[3-(4-Methylphenyl)-1,2,4-oxadiazol-5-yl]methyl}amine hydrochloride
{[3-(4-Methylphenyl)-1,2,4-oxadiazol-5-yl]methyl}amine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₂ClN₃O
MW: 225.68 g/mol
MDL Number: MFCD13186189
CAS Number: 890324-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR