[3-(Trifluoromethyl)-1,2,4-oxadiazol-5-yl]methanol

Supplier: Matrix Scientific

077259-500MG 077259-1G 077259-5G
102665-270EA 615.38 USD
102665-270 102665-272 102665-274
[3-(Trifluoromethyl)-1,2,4-oxadiazol-5-yl]methanol
[3-(Trifluoromethyl)-1,2,4-oxadiazol-5-yl]methanol

Hazard: Irritant

Formula: C₄H₃F₃N₂O₂
MW: 168.08 g/mol
MDL Number: MFCD20441335
CAS Number: 1338495-26-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR