(3-Methyl-1,2,4-oxadiazol-5-yl)methyl acetate

Supplier: Matrix Scientific

077258-500MG 077258-1G 077258-5G
102665-264EA 646.15 USD
102665-264 102665-266 102665-268
(3-Methyl-1,2,4-oxadiazol-5-yl)methyl acetate
(3-Methyl-1,2,4-oxadiazol-5-yl)methyl acetate

Hazard: Irritant

Formula: C₆H₈N₂O₃
MDL Number: MFCD11111647
CAS Number: 187970-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR