(2Z)-2-Cyano-2-(hydroxyimino)acetamide
Supplier: Matrix Scientific
077255-500MG
077255-1G
102665-248EA
646.15
USD
102665-248
102665-250
(2Z)-2-Cyano-2-(hydroxyimino)acetamide
(2Z)-2-Cyano-2-(hydroxyimino)acetamide
Hazard: Irritant
Formula:
C₃H₃N₃O₂ |
MDL Number:
MFCD03731348 CAS Number: 3849-20-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...