Methyl-2-methylpropionimidate hydrochloride

Supplier: Matrix Scientific

077254-500MG 077254-1G 077254-5G
102665-242EA 646.15 USD
102665-242 102665-244 102665-246
Methyl-2-methylpropionimidate hydrochloride
Methyl-2-methylpropionimidate hydrochloride

Hazard: Irritant

Formula: C₅H₁₂ClNO
MDL Number: MFCD03844704
CAS Number: 39739-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR