2-Chloro-5-methyl-1,3,4-thiadiazole

Supplier: Matrix Scientific

077252-500MG 077252-1G
102665-234EA 516.92 USD
102665-234 102665-236
2-Chloro-5-methyl-1,3,4-thiadiazole
2-Chloro-5-methyl-1,3,4-thiadiazole

Hazard: Irritant

Formula: C₃H₃ClN₂S
MDL Number: MFCD17170011
CAS Number: 53645-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR