Methyl-4-methyl-1H-imidazole-5-carboxylate

Supplier: Matrix Scientific

077250-500MG 077250-1G 077250-5G
102665-222EA 516.92 USD
102665-222 102665-224 102665-226
Methyl-4-methyl-1H-imidazole-5-carboxylate
Methyl-4-methyl-1H-imidazole-5-carboxylate

Hazard: Irritant

Formula: C₆H₈N₂O₂
MDL Number: MFCD06203737
CAS Number: 78892-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR