2-Chloro-5-ethyl-1,3,4-thiadiazole

Supplier: Matrix Scientific

077249-500MG 077249-1G
102665-218EA 516.92 USD
102665-218 102665-220
2-Chloro-5-ethyl-1,3,4-thiadiazole
2-Chloro-5-ethyl-1,3,4-thiadiazole

Hazard: Irritant

Formula: C₄H₅ClN₂S
MDL Number: MFCD17011731
CAS Number: 71859-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR