2-(2-Methyl-1H-imidazol-1-yl)ethanol hydrobromide
Supplier: Matrix Scientific
077248-500MG
077248-1G
102665-214EA
646.15
USD
102665-214
102665-216
2-(2-Methyl-1H-imidazol-1-yl)ethanol hydrobromide
2-(2-Methyl-1H-imidazol-1-yl)ethanol hydrobromide
Hazard: Irritant
Formula:
C₆H₁₁BrN₂O |
MDL Number:
MFCD20441423 CAS Number: 1615-15-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...