5-(Phenoxymethyl)-1,3,4-oxadiazol-2-amine

Supplier: Matrix Scientific

077247-500MG 077247-1G
102665-210EA 258.46 USD
102665-210 102665-212
5-(Phenoxymethyl)-1,3,4-oxadiazol-2-amine
5-(Phenoxymethyl)-1,3,4-oxadiazol-2-amine

Hazard: Irritant

Formula: C₉H₉N₃O₂
MDL Number: MFCD01710635
CAS Number: 21520-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR