3-Phenyl-1H-1,2,4-triazole

Supplier: Matrix Scientific

077244-500MG 077244-1G 077244-5G
102665-200EA 492.31 USD
102665-200 102665-202 102665-204
3-Phenyl-1H-1,2,4-triazole
3-Phenyl-1H-1,2,4-triazole

Hazard: Irritant

Formula: C₈H₇N₃
MDL Number: MFCD00159717
CAS Number: 3357-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR