4-(Chloromethyl)-1-methylpiperidine hydrochloride
Supplier: Matrix Scientific
077243-500MG
077243-1G
077243-5G
102665-194EA
58.46
USD
102665-194
102665-196
102665-198
4-(Chloromethyl)-1-methylpiperidine hydrochloride
4-(Chloromethyl)-1-methylpiperidine hydrochloride
Hazard: Irritant
Formula:
C₇H₁₅Cl₂N |
MDL Number:
MFCD12923013 CAS Number: 1182284-45-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...