5-tert-Butyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077241-500MG 077241-1G
102665-186EA 387.69 USD
102665-186 102665-188
5-tert-Butyl-4H-1,2,4-triazole-3-thiol
5-tert-Butyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₆H₁₁N₃S
MDL Number: MFCD00177195
CAS Number: 38449-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR