3-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)propanenitrile

Supplier: Matrix Scientific

Synonyms: 3-(1,3-Dioxoisoindolin-2-yl)propanenitrile

077240-500MG 077240-1G 077240-5G
102665-180EA 193.85 USD
102665-180 102665-182 102665-184
3-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)propanenitrile
3-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)propanenitrile

Hazard: Irritant

Formula: C₁₁H₈N₂O₂
MW: 200.2 g/mol
MDL Number: MFCD00090743
CAS Number: 3589-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR