5-Ethyl-1,3,4-thiadiazol-2-ol

Supplier: Matrix Scientific

077238-500MG 077238-1G
102665-172EA 516.92 USD
102665-172 102665-174
5-Ethyl-1,3,4-thiadiazol-2-ol
5-Ethyl-1,3,4-thiadiazol-2-ol

Hazard: Irritant

Formula: C₄H₆N₂OS
MDL Number: MFCD19982735
CAS Number: 1330756-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR