2-(Chloromethyl)-5-methoxy-1,3,4-thiadiazole

Supplier: Matrix Scientific

077237-500MG 077237-1G
102665-168EA 646.15 USD
102665-168 102665-170
2-(Chloromethyl)-5-methoxy-1,3,4-thiadiazole
2-(Chloromethyl)-5-methoxy-1,3,4-thiadiazole

Hazard: Irritant

Formula: C₄H₅ClN₂OS
MDL Number: MFCD12913232
CAS Number: 3914-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR