2-(Bromomethyl)-1-methylpiperidine hydrobromide

Supplier: Matrix Scientific

077236-500MG 077236-1G 077236-5G
102665-162EA 600 USD
102665-162 102665-164 102665-166
2-(Bromomethyl)-1-methylpiperidine hydrobromide
2-(Bromomethyl)-1-methylpiperidine hydrobromide

Hazard: Irritant

Formula: C₇H₁₅Br₂N
MDL Number: MFCD00179635
CAS Number: 1011407-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR