3-(Bromomethyl)-1-methylpyrrolidine hydrobromide

Supplier: Matrix Scientific

077235-500MG 077235-1G 077235-5G
102665-156EA 615.38 USD
102665-156 102665-158 102665-160
3-(Bromomethyl)-1-methylpyrrolidine hydrobromide
3-(Bromomethyl)-1-methylpyrrolidine hydrobromide

Hazard: Irritant

Formula: C₆H₁₃Br₂N
MDL Number: MFCD20441434
CAS Number: 1390654-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR