N-(5-Bromo-4-methyl-1,3-thiazol-2-yl)acetamide

Supplier: Matrix Scientific

077230-500MG 077230-1G
102665-134EA 387.69 USD
102665-134 102665-136
N-(5-Bromo-4-methyl-1,3-thiazol-2-yl)acetamide
N-(5-Bromo-4-methyl-1,3-thiazol-2-yl)acetamide

Hazard: Irritant

Formula: C₆H₇BrN₂OS
MDL Number: MFCD01924770
CAS Number: 21478-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR