1,2-Dimethylhydrazine
Supplier: Matrix Scientific
077227-500MG
077227-1G
077227-5G
102665-118EA
160
USD
102665-118
102665-120
102665-122
1,2-Dimethylhydrazine
1,2-Dimethylhydrazine
Hazard: Flammable/Toxic by inhalation
Formula:
C₂H₈N₂ Boiling Pt: 87 °C Melting Pt: –9 °C Density: 0.83 |
MDL Number:
MFCD01741277 CAS Number: 540-73-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...