N'-Benzoyl-N,N'-dimethylbenzohydrazide

Supplier: Matrix Scientific

077224-500MG 077224-1G 077224-5G
102665-102EA 161.54 USD
102665-102 102665-104 102665-106
N'-Benzoyl-N,N'-dimethylbenzohydrazide
N'-Benzoyl-N,N'-dimethylbenzohydrazide

Hazard: Irritant

Formula: C₁₆H₁₆N₂O₂
MDL Number: MFCD00020672
CAS Number: 1226-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR