3-(Chloromethyl)-1-ethylpyrrolidine hydrochloride

Supplier: Matrix Scientific

077217-500MG 077217-1G
102665-068EA 387.69 USD
102665-068 102665-070
3-(Chloromethyl)-1-ethylpyrrolidine hydrochloride
3-(Chloromethyl)-1-ethylpyrrolidine hydrochloride

Hazard: Irritant

Formula: C₇H₁₅Cl₂N
MDL Number: MFCD20441424
CAS Number: 98338-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR