N-(4-Methyl-2-nitrophenyl)acetamide

Supplier: Matrix Scientific

077215-500MG 077215-1G
102665-064EA 387.69 USD
102665-064 102665-066
N-(4-Methyl-2-nitrophenyl)acetamide
N-(4-Methyl-2-nitrophenyl)acetamide

Hazard: Irritant

Formula: C₉H₁₀N₂O₃
MDL Number: MFCD00024538
CAS Number: 612-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR